Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD3 Rabbit anti-Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89127896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-896 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-896 Supplier Abnova Corporation Supplier No. PAB26255
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FZD3.

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development. [provided by RefSeq

Specifications

Antigen FZD3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FZD3.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene FZD3
Gene Accession No. Q9NPG1
Gene Alias Fz-3/hFz3
Gene Symbols FZD3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acid at N-terminus of human FZD3.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7976
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except FZD6 (39%).
Target Species Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.