Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD5 Rabbit anti-Bovine, Dog, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-234 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-234 Supplier Abnova Corporation Supplier No. PAB16363
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of FZD5.

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand. [provided by RefSeq]

Specifications

Antigen FZD5
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of FZD5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene FZD5
Gene Accession No. Q13467
Gene Alias C2orf31/DKFZp434E2135/HFZ5/MGC129692
Gene Symbols FZD5
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human FZD5.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7855
Test Specificity N-terminal extracellular domain of human.
Target Species Bovine, Canine, Human, Monkey, Pig, Rabbit
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.