Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabbr1 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117240 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-240 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-240 Supplier Abnova Corporation Supplier No. PAB14687
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Gabbr1.

Specifications

Antigen Gabbr1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Gabbr1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Gabbr1
Gene Alias GABAbR1/bM573K1.1
Gene Symbols Gabbr1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with THG) corresponding to amino acids 100-124 of mouse Gabb1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54393
Test Specificity This antibody recognizes N-terminus of mouse gamma-aminobutyric acid (GABA) B receptor 1a. GB1a apparent MW ∼130 KDa.
Target Species Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.