Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabbr1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117924 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-924 Supplier Abnova Corporation Supplier No. PAB15872

Rabbit polyclonal antibody raised against synthetic peptide of Gabbr1.

Sequence: PHLPRPHPRVPPHPSSERRAVY

Specifications

Antigen Gabbr1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description gamma-aminobutyric acid (GABA) B receptor 1
Formulation In PBS (0.02% sodium azide)
Gene Gabbr1
Gene Symbols Gabbr1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 33-54 of rat Gabbr1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81657
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.