Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABBR1 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89127859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-859 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-859 Supplier Abnova Corporation Supplier No. PAB26219
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GABBR1.

Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the mammalian central nervous system. GABA exerts its effects through ionotropic [GABA(A/C)] receptors, to produce fast synaptic inhibition, and metabotropic [GABA(B)] receptors, to produce slow, prolonged inhibitory signals. The GABA(B) receptor consists of a heterodimer of two related 7-transmembrane receptors, GABA(B) receptor 1 and GABA(B) receptor 2. The GABA(B) receptor 1 gene is mapped to chromosome 6p21.3 within the HLA class I region close to the HLA-F gene. Susceptibility loci for multiple sclerosis, epilepsy, and schizophrenia have also been mapped in this region. Alternative splicing of this gene generates multiple transcript variants. [provided by RefSeq

Specifications

Antigen GABBR1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GABBR1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7-13 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GABBR1
Gene Accession No. NP_001461.1
Gene Alias FLJ92613/GABAB(1e)/GABABR1/GABBR1-3/GPRC3A/dJ271M21.1.1/dJ271M21.1.2/hGB1a
Gene Symbols GABBR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acid at N-terminus of human GABBR1.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2550
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Bovine, Canine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.