Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabbr2 (phospho S783), Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117121 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-121 Supplier Abnova Corporation Supplier No. PAB14529
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gabbr2.

Specifications

Antigen Gabbr2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gabbr2.
Dilution Western Blot (1:1000) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Gabbr2
Gene Accession No. O88871
Gene Alias Gpr51
Gene Symbols Gabbr2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S783 of rat Gabbr2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83633
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.