Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabrg2 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117128 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-128 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-128 Supplier Abnova Corporation Supplier No. PAB14537
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Gabrg2.

Specifications

Antigen Gabrg2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Gabrg2.
Dilution Western Blot (1:1000) Immunohistochemistry (1:400) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Gabrg2
Gene Accession No. P18508
Gene Symbols Gabrg2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to rat Gabrg2.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29709
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.