Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAL11, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115216 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-216 Supplier Abnova Corporation Supplier No. PAB1161

Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae GAL11.

Specifications

Antigen GAL11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae GAL11.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene GAL11
Gene Alias ABE1/MED15/RAR3/SDS4/SPT13
Gene Symbols GAL11
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-300 Saccharomyces cerevisiae GAL11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854106
Target Species Yeast
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.