Learn More
GAP43 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GAP43.
Supplier: Abnova Corporation PAB7977
Description
The protein encoded by this gene has been termed a 'growth' or 'plasticity' protein because it is expressed at high levels in neuronal growth cones during development and axonal regeneration. This protein is considered a crucial component of an effective regenerative response in the nervous system. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Specifications
GAP43 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GAP43. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
B-50/PP46 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2596 | |
Human, Mouse, Rat | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
GAP43 | |
GAP43 | |
A synthetic peptide corresponding to C-terminus of human GAP43. | |
100 μg | |
Primary | |
Identical to the related rat and mouse sequence. | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only