Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gap43 (phospho S41), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-306 Supplier Abnova Corporation Supplier No. PAB9654
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gap43.

Specifications

Antigen Gap43
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gap43.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Gap43
Gene Accession No. P07936
Gene Alias Basp2/MGC156666
Gene Symbols Gap43
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S41 of rat Gap43.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29423
Test Specificity Specific for the ∼50k Gap43 protein phosphorylated at Ser41. In some tissues This antibody also recognizes a higher molecular weight protein that is also recognized by the pan Gap43 antibody, that may be a Gap43 aggregate or oligomer.
Target Species Bovine, Canine, Chicken, Fish, Frog, Human, Mouse, Primate, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.