Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gap43 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115599 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-599 Supplier Abnova Corporation Supplier No. PAB12102
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Gap43.

Sequence: KEDPEADQEHA

Specifications

Antigen Gap43
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Gap43.
Dilution Western Blot (0.1-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by
Formulation In PBS (0.05% sodium azide)
Gene Accession No. P07936
Gene Alias B-50/Basp2/GAP-43
Gene Symbols Gap43
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 216-226 of mouse/rat Gap43.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14432/29423
Test Specificity This antibody is specific to GAP-43.
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.