Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GART, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130085 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-085 Supplier Abnova Corporation Supplier No. PAB28573
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant GART.

The protein encoded by this gene is a trifunctional polypeptide. It has phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase activity which is required for de novo purine biosynthesis. This enzyme is highly conserved in vertebrates. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: IEFVVVGPEAPLAAGIVGNLRSAGVQCFGPTAEAAQLESSKRFAKEFMDRHGIPTAQWKAFTKPEEACSFILSADFPALVVKASGLAAGKGVIVAKSKEEACKAVQEIMQEKAFGAAGETIVIEELLDGEEVSCLCFTDGKTV

Specifications

Antigen GART
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant GART.
Dilution Western Blot (Human: 1:100-1:250, Mouse/Rat: 1:100-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene GART
Gene Accession No. C9JBJ1
Gene Alias AIRS/GARS/GARTF/MGC47764/PAIS/PGFT/PRGS
Gene Symbols GART
Host Species Rabbit
Immunogen Recombinant protein corresponding to human GART.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2618
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.