Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBAS Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118511 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-511 Supplier Abnova Corporation Supplier No. PAB16676
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GBAS.

Chromosomal region 7p12, which contains GBAS, is amplified in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumor.The predicted 286-amino acid protein contains a signal peptide, a transmembrane domain, and 2 tyrosine phosphorylation sites. The GBAS transcript is expressed most abundantly in heart and skeletal muscle. GBAS protein might be involved in vesicular transport. [provided by RefSeq

Specifications

Antigen GBAS
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GBAS.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene GBAS
Gene Accession No. O75323
Gene Alias NIPSNAP2
Gene Symbols GBAS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 14 amino acids of human GBAS.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2631
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.