Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCGR Rabbit anti-Bovine, Human, Monkey, Polyclonal Antibody, Abnova™

Catalog No. 89127978 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-978 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-978 Supplier Abnova Corporation Supplier No. PAB26337
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GCGR.

The physiologic effects of glucagon (GCG; MIM 138030) are mediated through the glucagon receptor, a member of the superfamily of receptors characterized by a 7-transmembrane domain structure and by their coupling via GTP-binding proteins (G proteins) to adenyl cyclase.[supplied by OMIM

Specifications

Antigen GCGR
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GCGR.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (8 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GCGR
Gene Accession No. P47871
Gene Alias GGR/MGC138246
Gene Symbols GCGR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 20 amino acids at N-terminal extracellular domain of human GCGR.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2642
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Bovine, Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.