Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCNT6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GCNT6.
Supplier: Abnova Corporation PAB17629
Specifications
GCNT6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GCNT6. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q5T4J0 | |
GCNT6 | |
A synthetic peptide corresponding to internal of human GCNT6. | |
RUO | |
644378 | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
GCNT6 | |
bA421M1.3 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total GCNT6 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction