Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCOM1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120955 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-955 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-955 Supplier Abnova Corporation Supplier No. PAB19420
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GCOM1.

This gene (Gcom1) is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts. The GRINL1A gene uses a downstream promoter for transcription and also has multiple alternatively spliced variants. [provided by RefSeq

Specifications

Antigen GCOM1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GCOM1.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene GCOM1
Gene Accession No. NP_001018110
Gene Alias FLJ30973/GRINL1A/Gcom2/Gup1/Gup2/MGC126694/MGC138353
Gene Symbols GCOM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids near C-terminus of human GCOM1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145781
Test Specificity Multiple isoforms of GCOM1 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.