Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEMIN5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124803 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-803 Supplier Abnova Corporation Supplier No. PAB23104
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant GEMIN5.

GEMIN5 is part of a large macromolecular complex localized to both the cytoplasm and the nucleus that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), and GEMIN4 (MIM 606969).[supplied by OMIM

Sequence: YDSGSFTIMQEVYSAFLPDGCDHLRDKLGDHQSPATPAFKSLEAFFLYGRLYEFWWSLSRPCPNSSVWVRAGHRTLSVEPSQQLDTASTEETDPET

Specifications

Antigen GEMIN5
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant GEMIN5.
Dilution Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene GEMIN5
Gene Accession No. Q8TEQ6
Gene Alias DKFZp586M1824/MGC142174
Gene Symbols GEMIN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human GEMIN5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25929
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.