Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gfra3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Gfra3.
Supplier: Abnova Corporation PAB9892
Specifications
Gfra3 | |
Polyclonal | |
glial cell line derived neurotrophic factor family receptor alpha 3 | |
Gfra3 | |
Gfra3 | |
A synthetic peptide corresponding to amino acids 347-360 of mouse Gfra3. | |
RUO | |
14587 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
Y15110 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction