Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GH1/GH2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-234 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-234 Supplier Abnova Corporation Supplier No. PAB14677

Rabbit polyclonal antibody raised against recombinant GH1/GH2.

Specifications

Antigen GH1/GH2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant GH1/GH2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4
Gene Alias GH/GH-N/GHN/hGH-N
Gene Symbols GH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human GH1/GH2.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2688/2689
Test Specificity This antibody recognizes human growth hormone.
Target Species Human
Content And Storage Store at -20°C..
After reconstitution with 1 mL of deionized water, store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.