Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHITM Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89119401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-401 Supplier Abnova Corporation Supplier No. PAB17736
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GHITM.

Specifications

Antigen GHITM
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GHITM.
Dilution Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene GHITM
Gene Accession No. Q9H3K2
Gene Alias DERP2/DKFZp566C0746/FLJ26584/HSPC282/MICS1/My021/PTD010/TMBIM5
Gene Symbols GHITM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human GHITM.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27069
Test Specificity This antibody detects endogenous levels of total GHITM protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.