Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89127960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-960 Supplier Abnova Corporation Supplier No. PAB26319
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GIPR.

Gastric inhibitory polypeptide (GIP; MIM 137240), also called glucose-dependent insulinotropic polypeptide, is a 42-amino acid polypeptide synthesized by K cells of the duodenum and small intestine. It was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release potently in the presence of elevated glucose. The insulinotropic effect on pancreatic islet beta-cells was then recognized to be the principal physiologic action of GIP. Together with glucagon-like peptide-1, GIP is largely responsible for the secretion of insulin after eating. It is involved in several other facets of the anabolic response.[supplied by OMIM

Specifications

Antigen GIPR
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GIPR.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7-17 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GIPR
Gene Accession No. P48546
Gene Alias MGC126722
Gene Symbols GIPR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at N-terminal extracellular domain of human GIPR.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2696
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except GHRHR (100%), ZNF839 (61%).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.