Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gjb2, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial synthetic peptide of human GJB2.

Supplier:  Abnova Corporation PAB7965

Catalog No. 89-133-534


Only null left
Add to Cart

Description

Description

This gene encodes a member of the gap junction protein family. The gap junctions were first characterized by electron microscopy as regionally specialized structures on plasma membranes of contacting adherent cells. These structures were shown to consist of cell-to-cell channels that facilitate the transfer of ions and small molecules between cells. The gap junction proteins, also known as connexins, purified from fractions of enriched gap junctions from different tissues differ. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene are responsible for as much as 50% of pre-lingual, recessive deafness. [provided by RefSeq

Sequence: MDWGTLQTILGGVNKH
Specifications

Specifications

Gjb2
Polyclonal
Rabbit polyclonal antibody raised against partial synthetic peptide of human GJB2.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
CX26/DFNA3/DFNB1/HID/KID/NSRD1/PPK
Rabbit
Affinity purification
RUO
2706
Human, Mouse, Rat
Lyophilized
Western Blot
Unconjugated
Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user.
GJB2
GJB2
A synthetic peptide corresponding to amino acids 1-16 of human GJB2.
100 μg
Primary
The synthetic peptide of this antibody is different from the related mouse and rat sequences by two amino acids.
Store at -20°C.
After reconstitution with 0.2 mL of distilled water and concentration will be 500 ug/mL, store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only