Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glu-Glu tag Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-902 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-902 Supplier Abnova Corporation Supplier No. PAB0881
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Glu-Glu tag.

Specifications

Antigen Glu-Glu tag
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Glu-Glu tag.
Dilution Western Blot (1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.09% sodium azide)
Host Species Rabbit
Immunogen Glu-Glu (EYMPME) from the Polyoma middle T protein.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with proteins containing the Glu-Glu epitope tag.
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.