Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138216 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-216 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-216 Supplier Abnova Corporation Supplier No. PAB16661
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Glucagon.

Specifications

Antigen Glucagon
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Glucagon.
Dilution Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (0.09% sodium azide)
Host Species Rabbit
Immunogen Native purified Glucagon from pig pancreatic (conjugated with BSA).
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Pig, Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.