Learn More
GOLIM4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GOLIM4
Supplier: Abnova Corporation PAB28477
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus. [provided by RefSeq
Sequence: EHLEEEHDPSPEEQDREWKEQHEQREAANLLEGHARAEVYPSAKPMIKFQSPYEEQLEQQRLAVQQVEEAQQLREHQEALHQQRLQGHLLRQQEQQQQQVAREMALQRQAELEEGRPQHQEQLRQQAHYDAMDNDIVQGAEDSpecifications
GOLIM4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant GOLIM4 | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
F8W785 | |
GOLIM4 | |
Recombinant protein corresponding to amino acids of human GOLIM4 | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry(1:200-1:500) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
GOLIM4 | |
GIMPC/GOLPH4/GPP130/P138 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
27333 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only