Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gpd1l, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-751 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-751 Supplier Abnova Corporation Supplier No. PAB15686
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Gpd1l.

Sequence: FKELLQTPNFRITVVDDADTVELCGALKNIVAVGAGFCDGLRCGDNTKAAVIRLGLMEMIAFAKIFCKGQVSTATFLESCGVADLITTCYGGRNRRVAEAFARTGKTIEELEKELLNGQKLQGPQTSAEVYRILRQKGLLDKFPLFTAVYQICYEGRPVTQMLSCLQSHPEHI

Specifications

Antigen Gpd1l
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Gpd1l.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Gpd1l
Gene Accession No. AK172886
Gene Alias 2210409H23Rik/D9Ertd660e
Gene Symbols Gpd1l
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 173 mouse Gpd1l.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333433
Test Specificity Specific to recombinant protein GX2087. This antibody detects endogenous mGPD1L protein in several cell types.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.