Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-GPR119, Polyclonal, Abnova™

Catalog No. 89127662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-662 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-662 Supplier Abnova Corporation Supplier No. PAB26024

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen GPR119
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Gene Accession No. Q8TDV5
Gene Symbols GPR119
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at internal region of human GPR119.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 139760
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.