Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR132 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-209 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-209 Supplier Abnova Corporation Supplier No. PAB16336
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR132.

This gene encodes a subfamily member of the G-protein couple receptor (GPCR) superfamily. The encoded protein is a high-affinity receptor for lysophosphatidylcholine (LPC), a major phospholipid component of oxidized low density lipoprotein. This protein may react to LPC levels at sites of inflammation to limit the expansion of tissue-infiltrating cells. A similar protein in mouse is involved in cell cycle progression. [provided by RefSeq

Specifications

Antigen GPR132
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR132.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (12 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR132
Gene Accession No. Q9UNW8
Gene Alias G2A/MGC99642
Gene Symbols GPR132
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human GPR132.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29933
Test Specificity C-terminal domain of human.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.