Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR153 Rabbit anti-Bovine, Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89127923 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-923 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-923 Supplier Abnova Corporation Supplier No. PAB26283
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR153.

Specifications

Antigen GPR153
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR153.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (12 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR153
Gene Accession No. NP_997253.1
Gene Alias DKFZp762B2210/PGR1
Gene Symbols GPR153
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at 2nd extracellular domain of human GPR153.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387509
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except GPR162 (53%).
Target Species Bovine, Canine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.