Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR3 Rabbit anti-Human, Mouse, Pig, Polyclonal Antibody, Abnova™

Catalog No. 89127914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-914 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-914 Supplier Abnova Corporation Supplier No. PAB26274
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR3.

Specifications

Antigen GPR3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR3.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR3
Gene Accession No. P46089
Gene Alias ACCA
Gene Symbols GPR3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at 1st extracellular domain of human GPR3.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2827
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except SPPL2A (60%).
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.