Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR44 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-930 Supplier Abnova Corporation Supplier No. PAB15878

Rabbit polyclonal antibody raised against recombinant GPR44.

G protein-coupled receptors (GPCRs), such as GPR44, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.[supplied by OMIM

Specifications

Antigen GPR44
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description G protein-coupled receptor 44
Formulation In PBS (0.02% sodium azide)
Gene GPR44
Gene Alias CD294, CRTH2, DP2
Gene Symbols GPR44
Host Species Rabbit
Immunogen Recombinant protein corresponding to human GPR44.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11251
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.