Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR65 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89128017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-017 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-017 Supplier Abnova Corporation Supplier No. PAB26377
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR65.

Specifications

Antigen GPR65
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR65.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-11 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR65
Gene Accession No. Q8IYL9
Gene Alias TDAG8/hTDAG8
Gene Symbols GPR65
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids at 1st cytoplasmic domain of human GPR65.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8477
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.