Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR75 Rabbit anti-Bovine, Hamster, Horse, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR75.
Supplier: Abnova Corporation PAB26455
Description
GPR75 is a member of the G protein-coupled receptor family. GPRs are cell surface receptors that activate guanine-nucleotide binding proteins upon the binding of a ligand.[supplied by OMIM
Specifications
| GPR75 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR75. | |
| In PBS (0.09% sodium azide) | |
| O95800 | |
| GPR75 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human GPR75. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (23 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR75 | |
| GPR-chr2/WI-31133 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 10936 | |
| Bovine, Equine, Hamster, Human, Monkey, Pig, Rabbit | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction