Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR77 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-235 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-235 Supplier Abnova Corporation Supplier No. PAB16364
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR77.

The anaphylatoxins C3a (see MIM 120700), C4a (see MIM 120810), and C5a are cationic fragments generated during the complement cascade that participate in host defense. In the case of inappropriate complement activation, anaphylatoxins may be involved in autoimmunity and sepsis. C5L2 is coexpressed with the C5a receptor, C5AR (C5R1; MIM 113995), on polymorphonuclear neutrophils and may modulate C5AR activity (Gerard et al., 2005 [PubMed 16204243]).[supplied by OMIM

Specifications

Antigen GPR77
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR77.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR77
Gene Accession No. Q9P296
Gene Alias C5L2/GPF77
Gene Symbols GPR77
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human GPR77.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27202
Test Specificity N-terminal extracellular domain of human.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.