Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR89A Rabbit anti-Bovine, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118120 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-120 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-120 Supplier Abnova Corporation Supplier No. PAB16236
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GPR89A.

GPR89A is a nearly identical copy of the GPR89B gene (MIM 612806).[supplied by OMIM

Specifications

Antigen GPR89A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR89A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR89A
Gene Accession No. Q9Y302
Gene Alias FLJ10492/FLJ11696/FLJ36909/MGC688/MGC87324/UNQ192
Gene Symbols GPR89A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human GPR89A.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653519
Test Specificity Cytoplasmic loop. Predicted crossreactivity with mouse due to sequence similarity .
Target Species Bovine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.