Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-GPR98, Polyclonal,

Catalog No. ABN1377 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1377 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1377 Supplier MilliporeSigma™ Supplier No. ABN1377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GPR98 in Human samples, and it is validated for Immunohistochemistry

G-protein coupled receptor 98 (UniProt Q8WXG9; also known as Monogenic audiogenic seizure susceptibility protein 1 homolog, Usher syndrome type-2C protein, Very large G-protein coupled receptor 1) is encoded by the GPR98 (also known as KIAA0686, KIAA1943, MASS1, USH2C, VLGR1) gene (Gene ID 84059) in human. GPR98, also known as Very Large GPCR receptor 1 (Vlg1R1), is the largest G-protein-coupled receptor (GPCR) known. It is an adhesion-GPCR family receptor expressed in the central nervous system. GPR98 is produced with a signal peptide (a.a. 1-29) sequence, the removal of which yields the mature seven-transmembrane (a.a. 5909-5929, 5940-5960, 5980-6000, 6011-6031, 6060-6080, 6105-6125, 6134-6154) protein with a C-termiinal cytoplasmic domain (a.a. 6155-6306) and a large N-terminal extracellular region (a.a. 30-5908) that contains six epilepsy-associated repeats or EAR domains (a.a. 3189-3534) sandwiched between two clusters of calcium-binding Calx-beta repeats (Calx-beta 1-22/a.a. 30-3172 and Calx-beta 23-35/a.a. 3525-5468), as well as a GPCR-proteolytic site (GPS; a.a. 5853-5902) proximal to the first transmembrane helix. The extreme carboxy-terminus contains a consensus PDZ ligand sequence, suggesting interactions with other cytosolic or cytoskeletal proteins. Although the GPR98 ligand is yet to be identified, GPR98 gene mutations are linked to one form (2C) of Usher syndrome, the most common genetic cause of combined blindness and deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR98
Applications Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8WXG9
Gene Symbols GPR98; KIAA0686; KIAA1943; MASS1; USH2C; VLGR1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence from the second Calx-beta repeat of human GPR98.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_115495
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.