Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Grainyhead-Like 3/GRHL3, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABD68
Description
Specifically detects Grainyhead-Like 3/GRHL3 in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting
Grainyhead-like protein 3 homolog (UniProt Q8TE85; also known as Sister of mammalian grainyhead, Transcription factor CP2-like 4) is encoded by the GRHL3 (also known as SOM, TFCP2L4, VWS2) gene (Gene ID 57822) in human. Mammalian grainyhead-like (GRHL1, GRHL2, and GRHL3) genes code for transcription factors and are named after their drosophila homolog grainyhead (GRH). These factors show tissue- and developmental stage-dependent expression patterns. Grainyhead-like 3 (GRHL3) is reported to play a protective role against squamous cell carcinoma (SCC) of the skin. Tumor suppressor phosphatase and tensin homolog (PTEN) is a direct target of GRHL3, Grhl3-knockdown in mice causes skin tumor formation as a result of reduced Pten expression. Consistently, downregulated GRHL3 and PTEN levels are found in human SCC skin samples. In addition, GRHL3 is a positive regulator of E-cadherin expression during mesenchymal epithelial transition (MET) in non-tumorigenic mammary gland cells, as well as in breast and lung cancer cells. High GRHL3 expression is found in early stages of breast cancer and the expression is greatly reduced in advanced stages of breast cancer.
Specifications
Grainyhead-Like 3/GRHL3 | |
Polyclonal | |
Q8TE85 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
GRHL3; SOM; TFCP2L4; VWS2 | |
KLH-conjugated linear peptide corresponding to an internal sequence from the C-terminal half of human Grainyhead-Like 3/GRHL3. | |
100 μg | |
Stem Cell Research | |
NP_937817 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction