Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Grasp Rabbit anti-Bovine, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Grasp.
Supplier: Abnova Corporation PAB9619
Specifications
Grasp | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Grasp. | |
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
Q9JHZ4 | |
Rabbit | |
Affinity purification | |
RUO | |
192254 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Grasp | |
Grasp | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus rat Grasp. | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction