Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-GRASP55, Polyclonal,

Catalog No. ABS1520MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1520MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1520MI Supplier MilliporeSigma™ Supplier No. ABS1520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GRASP55 in Hamster, Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Golgi reassembly stacking protein of 55kDa (also called Golgi reassembly-stacking protein of 55kDa, GRASP55, or Grs2) is encoded by the gene designated as Gorasp2 in rat & mouse (GORASP2/GRS2/GOLPH6 in human). Golgi stack formation involves two homologous peripheral Golgi proteins, GRASP55 and GRASP65, localized to the medial-trans and cis cisternae, respectively. GRASP55 and GRASP65 stack mammalian Golgi cisternae via a common mechanism of phosphorylation-regulated oligomerization. GRASP55 stacks Golgi membranes by forming oligomers through its N-terminal GRASP domain. This process is regulated by phosphorylation within the C-terminal serine/proline-rich domain. Expression of nonphosphorylatable GRASP55 mutants enhances Golgi stacking in interphase cells and inhibits Golgi disassembly during mitosis. Knockdown of either GRASP55 or GRASP65 by siRNA reduces the number of cisternae per Golgi stack, while simultaneous knockdown of both GRASP proteins leads to disassembly of the entire stack. (PMID 20083603). Anti-GRASP55, is a rabbit polyclonal antiserum raised against a recombinant immunogen encompassing region within the C-terminal half of rat GRASP55.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP55
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q9R064
Gene Symbols Gorasp2
Host Species Rabbit
Immunogen Recombinant protein corresponding to the C-terminal of rat GRASP55.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) AF110267
Target Species Hamster, Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.