Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-GRASP65, Polyclonal,

Catalog No. ABS1601MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1601MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1601MI Supplier MilliporeSigma™ Supplier No. ABS1601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GRASP65 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Golgi reassembly stacking protein of 65kDa (also called Golgi reassembly and stacking protein 1 or Golgi peripheral membrane protein p65) is encoded by the Grasp65 or Gorasp65 gene in rat (GRASP65, GORASP1, or GOLPH5 gene in human). It is is a Golgi structural protein and mediates cisternae stacking via self-aggregation as well as interactions with other Golgi membrane-docking proteins, such as GM130. GRASP65 dimers are shown to aggregate in vitro and form extensive trans oligomers, which disaggregate upon phosphorylation by cdc2-cyclin B and polo-like kinases, and re-aggregate upon dephosphorylation (PMID 12839990). Fluorescent imaging studies reveal a dynamic exchange between the membrane-associated and cytosolic GRASP65. (PMID 11706049).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP65
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Accession No. O35254
Gene Symbols Gorasp1; Grasp65
Host Species Rabbit
Immunogen GST-tagged Recombinant protein corresponding to rat GRASP65.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_062258
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.