Learn More
GRB14, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRB14.
Supplier: Abnova Corporation PAB17735
Description
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date. [provided by RefSeq
Specifications
GRB14 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GRB14. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q14449 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total GRB14 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
GRB14 | |
GRB14 | |
A synthetic peptide corresponding to internal of human GRB14. | |
RUO | |
2888 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only