Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grin1 Rabbit anti-Bovine, Dog, Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89139139 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-139 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-139 Supplier Abnova Corporation Supplier No. PAB9706
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Grin1.

Specifications

Antigen Grin1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Grin1.
Dilution Immunohistochemistry (Frozen sections) (1:1000-2000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 5mM ammonium bicarbonate
Gene Accession No. P35439
Host Species Rabbit
Immunogen A synthetic peptide corresponding to mouse and rat Grin1.
Purification Method Affinity purification
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Primate, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with PBS store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.