Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIN2B Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89133555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-555 Supplier Abnova Corporation Supplier No. PAB7997
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GRIN2B.

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq

Specifications

Antigen GRIN2B
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GRIN2B.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) Immunocytochemistry (0.5-1 ug/mL) The optimal working dilution should be determine
Formulation Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Gene GRIN2B
Gene Alias MGC142178/MGC142180/NMDAR2B/NR2B/hNR3
Gene Symbols GRIN2B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at N-terminus of human GRIN2B.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2904
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C. After reconstitution with deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.