Learn More
GRIN2C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRIN2C.
Supplier: Abnova Corporation PAB25173
Description
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D).
Specifications
GRIN2C | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
GRIN2C | |
GRIN2C | |
A synthetic peptide corresponding to GRIN2C. | |
100 μL | |
Primary | |
GRIN2C polyclonal antibody detects endogenous levels of GRIN2C protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of GRIN2C. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
NMDAR2C/NR2C | |
Rabbit | |
Affinity purification | |
RUO | |
2905 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only