Learn More
GRK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRK1.
Supplier: Abnova Corporation PAB4515
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates rhodopsin and initiates its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). [provided by RefSeq
Specifications
GRK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GRK1. | |
In PBS (0.09% sodium azide) | |
Q15835 | |
GRK1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GRK1. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
GRK1 | |
GPRK1/RHOK/RK | |
Rabbit | |
Protein G purification | |
RUO | |
6011 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only