Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grm1/Grm5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134275 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-275 Supplier Abnova Corporation Supplier No. PAB9616
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Grm1/Grm5.

Specifications

Antigen Grm1/Grm5
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Grm1/Grm5.
Dilution Immunohistochemistry (Frozen sections) (1:500) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Accession No. P31424
Gene Alias Gprc1a
Gene Symbols Grm1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus rat Grm1/Grm5.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24414/24418
Test Specificity Specific for the ∼125KDa monomer and the ∼250KDa dimers of mGluR5 and mGluR1.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.