Learn More
GRM5 Rabbit anti-Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRM5.
Supplier: Abnova Corporation PAB16484
Description
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
GRM5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GRM5. | |
In PBS (0.09% sodium azide) | |
P41594 | |
GRM5 | |
A synthetic peptide (conjugated with KLH) corresponding to human GRM5. | |
50 μg | |
Primary | |
N-terminal extracellular domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5.6 ug/mL) The optimal working dilution should be determined by the end user. | |
GRM5 | |
GPRC1E/MGLUR5/mGlu5 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2915 | |
Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only