Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRPR Rabbit anti-Bovine, Dog, Hamster, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89128249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-249 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-249 Supplier Abnova Corporation Supplier No. PAB26610
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of GRPR.

Gastrin-releasing peptide (GRP) regulates numerous functions of the gastrointestinal and central nervous systems, including release of gastrointestinal hormones, smooth muscle cell contraction, and epithelial cell proliferation and is a potent mitogen for neoplastic tissues. The effects of GRP are mediated through the gastrin-releasing peptide receptor. This receptor is a glycosylated, 7-transmembrane G-protein coupled receptor that activates the phospholipase C signaling pathway. The receptor is aberrantly expressed in numerous cancers such as those of the lung, colon, and prostate. An individual with autism and multiple exostoses was found to have a balanced translocation between chromosome 8 and a chromosome X breakpoint located within the gastrin-releasing peptide receptor gene. [provided by RefSeq

Specifications

Antigen GRPR
Applications Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GRPR.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GRPR
Gene Accession No. P30550
Gene Symbols GRPR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids from 3rd cytoplasmic domain of human GRPR.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2925
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except ADAM28 (41%).
Target Species Bovine, Canine, Hamster, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.