Learn More
GSC2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GSC2.
Supplier: Abnova Corporation PAB17336
Description
Goosecoidlike (GSCL), a homeodomain-containing gene, resides in the critical region for VCFS/DGS on 22q11. Velocardiofacial syndrome (VCFS) is a developmental disorder characterized by conotruncal heart defects, craniofacial anomalies, and learning disabilities. VCFS is phenotypically related to DiGeorge syndrome (DGS) and both syndromes are associated with hemizygous 22q11 deletions. Because many of the tissues and structures affected in VCFS/DGS derive from the pharyngeal arches of the developing embryo, it is believed that haploinsufficiency of a gene involved in embryonic development may be responsible for its etiology. The gene is expressed in a limited number of adult tissues, as well as in early human development. [provided by RefSeq
Specifications
GSC2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GSC2. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
O15499 | |
GSC2 | |
A synthetic peptide corresponding to internal of human GSC2. | |
RUO | |
2928 | |
Human, Mouse | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
GSC2 | |
GSCL | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total GSC2 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only