Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GST protein, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against GST protein.
Supplier: Abnova Corporation PAB1625E01P
Description
Sequence: MESPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLEDPGYRGRTSFV*Specifications
| GST protein | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against GST protein. | |
| U78874.1 | |
| GST protein. ( 242 a.a protein modified from AAB37352, please see the protein seq for the actual immunogen sequence. ) | |
| RUO | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction